siRNA Details
Gene Symbol: Tmem186 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025756.1 | NP_001020927.1 | 2 | 720 | ||
GenBank mRNA | BC098933.1 | - | 2 | 720 | 7386881 |
Gene Symbol: LOC100911247 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003750323.1 | XP_003750371.1 | 1 | 314 |
Gene Symbol: RGD1306282 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_235171.6 | XP_235171.6 | 2 | 590 |
