siRNA Details
Gene Symbol: LOC497899 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001017472.1 | NP_001017472.1 | 5 | 801 | ||
XM_006246312.1 | XP_006246374.1 | 5 | 869 | |||
XM_006246313.2 | XP_006246375.1 | 5 | 1444 | |||
XM_008767700.1 | XP_008765922.1 | 5 | 1332 | |||
XM_008767701.1 | XP_008765923.1 | 5 | 953 | |||
GenBank mRNA | BC079296.1 | - | 5 | 801 | 7134494 | |
FQ225097.1 | - | 5 | 836 |
