siRNA Details
Gene Symbol: Ccdc69 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109031.1 | NP_001102501.1 | 6 | 557 | ||
XM_006246393.2 | XP_006246455.1 | 7 | 663 | |||
XM_008767703.1 | XP_008765925.1 | 6 | 887 |

More Information
Other Species Targeted

