siRNA Details
Gene Symbol: LOC497934 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001017474.1 | NP_001017474.2 | 1 | 989 | ||
GenBank mRNA | BC082026.1 | - | 1 | 989 | 7129979 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_028005, AK009886, AK169875, AK151629.
