siRNA Details
Gene Symbol: LOC497940 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001017476.1 | NP_001017476.1 | 4 | 805 | ||
XM_006246777.2 | XP_006246839.1 | 4 | 952 | |||
XM_006246778.2 | XP_006246840.1 | 4 | 956 | |||
XM_006246781.2 | XP_006246843.1 | 4 | 953 | |||
XM_006246782.2 | XP_006246844.1 | 4 | 949 | |||
GenBank mRNA | BC079326.1 | - | 4 | 805 | 7135484 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
