siRNA Details
Gene Symbol: Gltpd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001115030.1 | NP_001108502.1 | 2 | 442 | ||
GenBank mRNA | BC091235.1 | - | 2 | 420 | ||
BC158886.1 | - | 2 | 442 | 8372488 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
