siRNA Details
Gene Symbol: Dph1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105809.2 | NP_001099279.1 | 13 | 1628 | ||
GenBank mRNA | BC166808.1 | - | 13 | 1628 | 7124384 | |
FQ228687.1 | - | 13 | 1644 |
Gene Symbol: Ovca2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109036.1 | NP_001102506.1 | 2 | 480 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
