siRNA Details
Gene Symbol: Tlcd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006246868.2 | XP_006246930.1 | 4 | 1342 | ||
XM_006246869.2 | XP_006246931.1 | 4, 5 | 1240 | |||
XM_573142.6 | XP_573142.4 | 4 | 1342 | |||
GenBank mRNA | FQ219135.1 | - | 4 | 814 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
