siRNA Details
Gene Symbol: Heatr9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006247018.2 | XP_006247080.1 | 4 | 1377 | ||
XM_006247020.2 | XP_006247082.1 | 4 | 1377 | |||
XM_006247021.2 | XP_006247083.1 | 3 | 1192 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
