siRNA Details
Gene Symbol: Heatr9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006247018.2 | XP_006247080.1 | 9 | 1913 | ||
XM_006247021.2 | XP_006247083.1 | 8 | 1728 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
