siRNA Details
Gene Symbol: Znhit3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_573169.6 | XP_573169.3 | 4, 5 | 308 | ||
GenBank mRNA | FQ231514.1 | - | 4, 5 | 285 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
