siRNA Details
Gene Symbol: Rad51c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001129777.1 | NP_001123249.1 | 3 | 495 | ||
XM_006247106.1 | XP_006247168.1 | 3 | 573 | |||
GenBank mRNA | BC166862.1 | - | 3 | 402 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_053269, BY720217, BC090648, AK076551, AK136204, and others.
Gene Ontology Categories:
