siRNA Details
Gene Symbol: Tmem100 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001017479.1 | NP_001017479.1 | 3 | 550 | ||
XM_008768100.1 | XP_008766322.1 | 2 | 1188 | |||
XM_008768101.1 | XP_008766323.1 | 3 | 1047 | |||
XM_008768102.1 | XP_008766324.1 | 3 | 777 | |||
GenBank mRNA | BC092577.1 | - | 3 | 550 | 7124708 | |
FQ212975.1 | - | 3 | 555 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
