siRNA Details
Gene Symbol: Hoxb7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001017480.1 | NP_001017480.1 | 2 | 606 | ||
GenBank mRNA | BC079340.1 | - | 2 | 606 | 7106610 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Hoxb7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001017480.1 | NP_001017480.1 | 2 | 606 | ||
GenBank mRNA | BC079340.1 | - | 2 | 606 | 7106610 |