siRNA Details
Gene Symbol: Hoxb5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191925.1 | NP_001178854.1 | 1 | 254 | ||
XM_008768104.1 | XP_008766326.1 | 1 | 2087 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_008268, BC103604, AK012146, BC103607, BC103595, and others.
Gene Ontology Categories:
