siRNA Details
Gene Symbol: Dusp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006247402.2 | XP_006247464.1 | 3 | 456 | ||
XM_006247403.2 | XP_006247465.1 | 3 | 455 | |||
GenBank mRNA | BC169043.1 | - | 2 | 325 | 9099479 |

More Information
Other Species Targeted

Gene Ontology Categories:
