siRNA Details
Gene Symbol: Dusp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006247402.2 | XP_006247464.1 | 3 | 453 | ||
XM_006247403.2 | XP_006247465.1 | 3 | 452 | |||
GenBank mRNA | BC169043.1 | - | 2 | 322 | 9099479 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
