siRNA Details
Gene Symbol: RGD1562667 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006247741.2 | XP_006247803.1 | 4 | 1178 | ||
XM_006247742.2 | XP_006247804.1 | 4 | 1093 | |||
XM_577143.5 | XP_577143.3 | 4 | 1323 |

More Information
Other Species Targeted

