siRNA Details
Gene Symbol: RGD1561778 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001168284.1 | NP_001161756.1 | 516 | |||
GenBank mRNA | GU138868.1 | - | 516 |
Gene Symbol: RGD1559482 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001168285.1 | NP_001161757.1 | 3 | 511 | ||
GenBank mRNA | GU138867.1 | - | 3 | 511 |
