siRNA Details
Gene Symbol: Fn3k | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109051.1 | NP_001102521.1 | 4 | 576 | ||
XM_006247907.2 | XP_006247969.1 | 6 | 1472 | |||
GenBank mRNA | FQ212712.1 | - | 8 | 1037 | ||
FQ212922.1 | - | 6 | 940 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
