siRNA Details
Gene Symbol: RGD1562726 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109052.1 | NP_001102522.1 | 3 | 719 | ||
XM_006248074.2 | XP_006248136.1 | 3 | 1022 | |||
XM_006248075.2 | XP_006248137.1 | 2 | 452 | |||
XM_006248076.2 | XP_006248138.1 | 3 | 535 | |||
XM_008768588.1 | XP_008766810.1 | 3 | 1001 | |||
XM_008768589.1 | XP_008766811.1 | 2 | 431 | |||
XM_008768590.1 | XP_008766812.1 | 4 | 2056 | |||
XM_008768591.1 | XP_008766813.1 | 3 | 514 | |||
XM_008768592.1 | XP_008766814.1 | 3 | 1473 | |||
XM_008768593.1 | XP_008766815.1 | 3 | 603 | |||
XM_008768594.1 | XP_008766816.1 | 4 | 1207 | |||
XM_008768595.1 | XP_008766817.1 | 4 | 1163 | |||
XM_008768596.1 | XP_008766818.1 | 2 | 790 | |||
XM_008768597.1 | XP_008766819.1 | 2 | 564 |

More Information
Other Species Targeted

