siRNA Details
Gene Symbol: Tmem127 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100978.1 | NP_001094448.1 | 3 | 572 | ||
GenBank mRNA | BC131850.1 | - | 3 | 572 | 7125680 | |
FQ230469.1 | - | 3 | 546 |

More Information
Other Species Targeted

Gene Ontology Categories:
