siRNA Details
Gene Symbol: Cd96 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025032.1 | NP_001020203.1 | 2 | 324 | ||
XM_008768647.1 | XP_008766869.1 | 2 | 519 | |||
GenBank mRNA | BC091206.1 | - | 2 | 324 | 7380788 | |
FQ228851.1 | - | 2 | 312 |
