siRNA Details
Gene Symbol: LOC100912534 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003751083.2 | XP_003751131.1 | 5 | 629 |
Gene Symbol: Polr2h | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134789.1 | NP_001128261.1 | 5 | 615 | ||
GenBank mRNA | FQ222889.1 | - | 5 | 613 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
