siRNA Details
Gene Symbol: Spidr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100988.1 | NP_001094458.1 | 3 | 238 | ||
XM_008768898.1 | XP_008767120.1 | 3 | 263 | |||
XM_008768899.1 | XP_008767121.1 | 3 | 263 | |||
GenBank mRNA | BC129120.1 | - | 3 | 238 | 7123928 |
