siRNA Details
Gene Symbol: LOC498122 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109058.1 | NP_001102528.1 | 2 | 114 | ||
XM_008768900.1 | XP_008767122.1 | 1 | 418 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
