siRNA Details
Gene Symbol: RGD1563425 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109059.1 | NP_001102529.1 | 6 | 1077 | ||
XM_008768988.1 | XP_008767210.1 | 7 | 1189 |

More Information
Other Species Targeted

