siRNA Details
Gene Symbol: N4bp2l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001035222.2 | NP_001030299.1 | 5 | 512 | ||
XM_008768992.1 | XP_008767214.1 | 5 | 512 | |||
GenBank mRNA | BC168658.1 | - | 5 | 512 | 7314496 |
