siRNA Details
Gene Symbol: Tex26 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134603.1 | NP_001128075.1 | 3 | 257 | ||
XM_006248811.2 | XP_006248873.1 | 3 | 396 | |||
XM_006248813.2 | XP_006248875.1 | 3 | 562 |
