siRNA Details
Gene Symbol: Gpr146 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109062.1 | NP_001102532.1 | 2 | 494 | ||
XM_006248955.2 | XP_006249017.1 | 2 | 2639 | |||
XM_006248956.2 | XP_006249018.1 | 2 | 973 | |||
XM_006248957.2 | XP_006249019.1 | 2 | 507 | |||
XM_006248958.2 | XP_006249020.1 | 3 | 2247 |
