siRNA Details
Gene Symbol: Gpr146 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109062.1 | NP_001102532.1 | 2 | 1011 | ||
XM_006248955.2 | XP_006249017.1 | 2 | 3156 | |||
XM_006248956.2 | XP_006249018.1 | 2 | 1490 | |||
XM_006248957.2 | XP_006249019.1 | 2 | 1024 | |||
XM_006248958.2 | XP_006249020.1 | 3 | 2764 |

More Information
Other Species Targeted

Set Membership: |
![]() |
Gene Ontology Categories:
