siRNA Details
Gene Symbol: LOC498154 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025033.1 | NP_001020204.1 | 4, 5 | 435 | ||
XM_006248959.2 | XP_006249021.1 | 3, 4 | 918 | |||
GenBank mRNA | BC088429.1 | - | 4, 5 | 435 | 7124272 |
