siRNA Details
Gene Symbol: MGC112692 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025759.1 | NP_001020930.1 | 669 | |||
XM_008768997.1 | XP_008767219.1 | 2 | 705 | |||
GenBank mRNA | BC081909.1 | - | 665 | |||
BC098706.1 | - | 669 | 7114401 |
