siRNA Details
Gene Symbol: Spry3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109063.1 | NP_001102533.1 | 2 | 440 | ||
XM_008769065.1 | XP_008767287.1 | 2 | 1814 | |||
XM_008769066.1 | XP_008767288.1 | 3 | 735 | |||
XM_008769067.1 | XP_008767289.1 | 3 | 687 | |||
XM_008769068.1 | XP_008767290.1 | 2 | 403 | |||
XM_008769069.1 | XP_008767291.1 | 2 | 533 | |||
GenBank mRNA | BC168971.1 | - | 2 | 433 | 7316797 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
