siRNA Details
Gene Symbol: Nxpe4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_001056892.5 | XP_001056892.3 | 2, 3 | 1057 | ||
XM_006249027.2 | XP_006249089.1 | 3, 4 | 911 | |||
XM_006249028.2 | XP_006249090.1 | 2, 3 | 1143 |
Gene Symbol: RGD1562319 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008769094.1 | XP_008767316.1 | 3, 4 | 964 |

More Information
Other Species Targeted

