siRNA Details
Gene Symbol: Bri3bp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001017487.1 | NP_001017487.1 | 2, 3 | 355 | ||
GenBank mRNA | BC085334.1 | - | 2, 3 | 355 | 7301029 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Bri3bp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001017487.1 | NP_001017487.1 | 2, 3 | 355 | ||
GenBank mRNA | BC085334.1 | - | 2, 3 | 355 | 7301029 |