siRNA Details
Gene Symbol: Mapkapk5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025761.2 | NP_001020932.2 | 6, 7 | 655 | ||
NM_001164043.1 | NP_001157515.1 | 9, 10 | 1024 | |||
GenBank mRNA | BC099161.1 | - | 6, 7 | 655 | 7381631 |
