siRNA Details
Gene Symbol: Iqcd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC083607.1 | - | 3, 4 | 1261 | 7133364 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Iqcd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC083607.1 | - | 3, 4 | 1261 | 7133364 |