siRNA Details
Gene Symbol: LOC498222 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100788.1 | NP_001094258.1 | 4 | 1424 | ||
XM_006249714.2 | XP_006249776.1 | 5 | 3686 | |||
XM_008769488.1 | XP_008767710.1 | 4 | 1549 | |||
XM_008769489.1 | XP_008767711.1 | 3 | 3394 | |||
GenBank mRNA | BC099745.1 | - | 1 | 680 |
