siRNA Details
Gene Symbol: Mfsd4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109069.1 | NP_001102539.1 | 6 | 1270 | ||
XM_006249791.2 | XP_006249853.1 | 6 | 1265 |

More Information
Other Species Targeted

Gene Ontology Categories:
