siRNA Details
Gene Symbol: Lgr6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_573455.6 | XP_573455.1 | 1 | 364 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Lgr6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_573455.6 | XP_573455.1 | 1 | 364 |