siRNA Details
Gene Symbol: LOC498236 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001047926.1 | NP_001041391.1 | 15 | 1826 | ||
XM_008769565.1 | XP_008767787.1 | 16 | 2025 | |||
XM_008769566.1 | XP_008767788.1 | 17 | 2094 | |||
XM_008769567.1 | XP_008767789.1 | 17 | 2094 | |||
GenBank mRNA | AY539937.1 | - | 15 | 1826 |
