siRNA Details
Gene Symbol: RGD1564614 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134792.1 | NP_001128264.1 | 5, 6 | 831 | ||
XM_006249945.2 | XP_006250007.1 | 6, 7 | 1054 | |||
XM_006249946.2 | XP_006250008.1 | 4, 5 | 682 | |||
XM_008769568.1 | XP_008767790.1 | 6, 7 | 1054 | |||
XM_008769569.1 | XP_008767791.1 | 6, 7 | 970 | |||
GenBank mRNA | AF436847.2 | - | 5, 6 | 831 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
