siRNA Details
Gene Symbol: Wdr26 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109081.1 | NP_001102551.1 | 10 | 1161 | ||
GenBank mRNA | FQ226610.1 | - | 1155 | |||
FQ227016.1 | - | 1155 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
