siRNA Details
Gene Symbol: Wdr26 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109081.1 | NP_001102551.1 | 5 | 533 | ||
GenBank mRNA | FQ226610.1 | - | 527 | |||
FQ227016.1 | - | 527 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_145514, AK082326, BC020044, BC139337, BC139336, and others.
