siRNA Details
Gene Symbol: LOC498308 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001047927.1 | NP_001041392.1 | 1, 2 | 89 | ||
XM_008769867.1 | XP_008768089.1 | 5, 6 | 1134 | |||
GenBank mRNA | AY325255.1 | - | 1, 2 | 89 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
