siRNA Details
Gene Symbol: Vash2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109082.1 | NP_001102552.1 | 6 | 961 | ||
XM_006250463.2 | XP_006250525.1 | 6 | 930 | |||
XM_006250465.2 | XP_006250527.1 | 6 | 930 | |||
XM_008769868.1 | XP_008768090.1 | 6 | 930 | |||
XM_008769869.1 | XP_008768091.1 | 6 | 1253 | |||
XM_008769870.1 | XP_008768092.1 | 6 | 930 | |||
XM_008769871.1 | XP_008768093.1 | 6 | 930 | |||
XM_008769872.1 | XP_008768094.1 | 6 | 930 | |||
GenBank mRNA | AY724511.1 | - | 6 | 875 |
