siRNA Details
Gene Symbol: LOC498330 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001017495.1 | NP_001017495.1 | 2 | 377 | ||
GenBank mRNA | BC079214.1 | - | 2 | 377 | 7131874 |

More Information
Other Species Targeted

