siRNA Details
Gene Symbol: LOC498339 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001047928.1 | NP_001041393.1 | 4 | 425 | ||
GenBank mRNA | AY539941.1 | - | 4 | 425 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
