siRNA Details
Gene Symbol: RGD1562136 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109086.1 | NP_001102556.1 | 3 | 675 | ||
GenBank mRNA | FQ210387.1 | - | 3 | 606 | ||
FQ218309.1 | - | 3 | 606 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
